Assay Details
| Gene Symbol: AIMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717177.3 | XP_002717223.1 | 3 - 4 | 346 | 128 | |
| GenBank mRNA | AJ879471.1 | - | 1 - 2 | 97 | 128 | |
More Information
Set Membership: |
|

AIMP1
Genomic Map