Assay Details
| Gene Symbol: FKBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718166.3 | XP_002718212.1 | 1 - 2 | 146 | 120 | |
| GenBank mRNA | D82876.1 | - | 1 - 2 | 104 | 120 | |
| Gene Symbol: LOC100348481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343517.1 | XP_017199006.1 | 1 | 132 | 120 | |
More Information
Set Membership: |
|

FKBP3
Genomic Map