Assay Details
| Gene Symbol: FKBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718166.3 | XP_002718212.1 | 5 - 6 | 560 | 109 | |
| GenBank mRNA | D82876.1 | - | 5 - 6 | 518 | 109 | |
More Information
Set Membership: |
|

FKBP3
Genomic Map