Assay Details
| Gene Symbol: RPLP0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719794.2 | XP_002719840.1 | 3 | 183 | 73 | |
| GenBank mRNA | DQ102477.1 | - | 1 | 46 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPLP0| Gene Symbol: RPLP0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719794.2 | XP_002719840.1 | 3 | 183 | 73 | |
| GenBank mRNA | DQ102477.1 | - | 1 | 46 | 73 | |
Set Membership: |
|