Assay Details
| Gene Symbol: LOC100009591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712249.3 | XP_002712295.1 | 7 - 8 | 1526 | 93 | |
| GenBank mRNA | DQ242485.1 | - | 1 - 2 | 127 | 93 | |
More Information
Set Membership: |
|

LOC100009591
Genomic Map