Assay Details
| Gene Symbol: LOC100009591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712249.3 | XP_002712295.1 | 9 - 10 | 1735 | 97 | |
| GenBank mRNA | DQ242485.1 | - | 3 - 4 | 336 | 97 | |
More Information
Set Membership: |
|

LOC100009591
Genomic Map