Assay Details
| Gene Symbol: ADCYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344061.1 | XP_017199550.1 | 4 - 5 | 722 | 84 | |
| GenBank mRNA | DQ421397.1 | - | 1 - 2 | 55 | 84 | |
More Information
Set Membership: |
|

ADCYAP1
Genomic Map