Assay Details
| Gene Symbol: CASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262043.2 | XP_008260265.1 | 5 - 6 | 659 | 80 | |
| XM_008262051.2 | XP_008260273.1 | 5 - 6 | 659 | 80 | ||
| XM_017343637.1 | XP_017199126.1 | 5 - 6 | 659 | 80 | ||
| GenBank mRNA | EF472882.1 | - | 1 - 2 | 151 | 80 | |
More Information
Set Membership: |
|

CASP1
Genomic Map