Assay Details
| Gene Symbol: BLMH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718903.3 | XP_002718949.1 | 7 - 8 | 1008 | 63 | |
| XM_008270953.2 | XP_008269175.1 | 8 - 9 | 1089 | 63 | ||
| XM_017349038.1 | XP_017204527.1 | 8 - 9 | 1092 | 63 | ||
| XM_017349039.1 | XP_017204528.1 | 8 - 9 | 1091 | 63 | ||
| GenBank mRNA | J02866.1 | - | 6 - 7 | 738 | 63 | |
More Information
Set Membership: |
|

BLMH
Genomic Map