Assay Details
| Gene Symbol: SSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712273.3 | XP_002712319.1 | 11 - 12 | 1257 | 123 | |
| GenBank mRNA | L08230.1 | - | 1 - 2 | 66 | 123 | |
More Information
Set Membership: |
|

SSB
Genomic Map