Assay Details
| Gene Symbol: PPP2R2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195718.1 | NP_001182647.1 | 5 - 6 | 708 | 64 | |
| GenBank mRNA | M64931.1 | - | 3 - 4 | 364 | 64 | |
More Information
Set Membership: |
|

PPP2R2B
Genomic Map