Assay Details
| Gene Symbol: ALOX5AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720606.3 | XP_002720652.1 | 264 | 61 | ||
| GenBank mRNA | M96556.1 | - | 175 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALOX5AP| Gene Symbol: ALOX5AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720606.3 | XP_002720652.1 | 264 | 61 | ||
| GenBank mRNA | M96556.1 | - | 175 | 61 | ||
Set Membership: |
|