Assay Details
| Gene Symbol: CA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195708.1 | NP_001182637.1 | 6 - 7 | 740 | 75 | |
| GenBank mRNA | EB382732.1 | - | 4 - 5 | 362 | 75 | |
| M98395.1 | - | 5 - 6 | 639 | 75 | ||
More Information
Set Membership: |
|

CA2
Genomic Map