Assay Details
| Gene Symbol: CNGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248606.2 | XP_008246828.1 | 6 - 7 | 574 | 149 | |
| GenBank mRNA | S65218.1 | - | 2 - 3 | 157 | 149 | |
More Information
Set Membership: |
|

CNGA1
Genomic Map