Assay Details
| Gene Symbol: AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195724.1 | NP_001182653.1 | 3 - 4 | 1823 | 83 | |
| GenBank mRNA | U16366.1 | - | 3 - 4 | 1247 | 83 | |
More Information
Set Membership: |
|

AR
Genomic Map