Assay Details
| Gene Symbol: CTSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721589.3 | XP_002721635.3 | 961 | 81 | ||
| GenBank mRNA | EF472892.1 | - | 570 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTSH| Gene Symbol: CTSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721589.3 | XP_002721635.3 | 961 | 81 | ||
| GenBank mRNA | EF472892.1 | - | 570 | 81 | ||
Set Membership: |
|