Assay Details
| Gene Symbol: CTSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715580.3 | XP_002715626.2 | 3 - 4 | 389 | 67 | |
| XM_017345904.1 | XP_017201393.1 | 3 - 4 | 389 | 67 | ||
| GenBank mRNA | EF472894.1 | - | 1 - 2 | 106 | 67 | |
More Information
Set Membership: |
|

CTSS
Genomic Map