Assay Details
| Gene Symbol: SLC16A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714902.3 | XP_002714948.1 | 5 - 6 | 1390 | 83 | |
| GenBank mRNA | EF489851.1 | - | 5 - 6 | 1040 | 83 | |
More Information
Set Membership: |
|

SLC16A10
Genomic Map