Assay Details
| Gene Symbol: CDK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711061.3 | XP_002711107.1 | 1 - 2 | 330 | 59 | |
| XM_002711062.3 | XP_002711108.1 | 1 - 2 | 331 | 59 | ||
| GenBank mRNA | EU137107.1 | - | 1 - 2 | 59 | 59 | |
More Information
Set Membership: |
|

CDK2
Genomic Map