Assay Details
| Gene Symbol: CCL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708013.3 | XP_002708059.1 | 3 - 4 | 446 | 133 | |
| GenBank mRNA | EU407478.1 | - | 2 - 3 | 304 | 133 | |
More Information
Set Membership: |
|

CCL21
Genomic Map