Assay Details
| Gene Symbol: MELTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081992.1 | NP_001075461.1 | 9 - 10 | 1293 | 54 | |
| GenBank mRNA | AB010995.1 | - | 9 - 10 | 1293 | 54 | |
More Information
Set Membership: |
|

MELTF
Genomic Map