Assay Details
| Gene Symbol: PLAU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082011.1 | NP_001075480.1 | 4 - 5 | 319 | 70 | |
| XM_008269903.2 | XP_008268125.1 | 4 - 5 | 242 | 70 | ||
| GenBank mRNA | AB087224.1 | - | 4 - 5 | 319 | 70 | |
| AY029517.1 | - | 4 - 5 | 311 | 70 | ||
| AY122285.1 | - | 4 - 5 | 260 | 70 | ||
More Information
Set Membership: |
|

PLAU
Genomic Map