Assay Details
| Gene Symbol: GYS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082023.1 | NP_001075492.1 | 800 | 62 | ||
| XM_017339799.1 | XP_017195288.1 | 794 | 62 | |||
| GenBank mRNA | AF017114.1 | - | 800 | 62 | ||
More Information
Set Membership: |
|

GYS1
Genomic Map