Assay Details
| Gene Symbol: LIPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082032.1 | NP_001075501.1 | 5 - 6 | 805 | 71 | |
| XM_017347897.1 | XP_017203386.1 | 5 - 6 | 743 | 71 | ||
| GenBank mRNA | AF041202.1 | - | 5 - 6 | 805 | 71 | |
More Information
Set Membership: |
|

LIPC
Genomic Map