Assay Details
| Gene Symbol: MMP13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082037.1 | NP_001075506.1 | 8 - 9 | 1877 | 71 | |
| GenBank mRNA | AF059201.1 | - | 8 - 9 | 1877 | 71 | |
More Information
Set Membership: |
|

MMP13
Genomic Map