Assay Details
| Gene Symbol: CNMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082040.1 | NP_001075509.1 | 5 - 6 | 788 | 61 | |
| GenBank mRNA | AF072129.1 | - | 5 - 6 | 788 | 61 | |
More Information
Set Membership: |
|

CNMD
Genomic Map