Assay Details
| Gene Symbol: IL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082045.1 | NP_001075514.1 | 1 - 2 | 165 | 70 | |
| XM_008268045.2 | XP_008266267.1 | 1 - 2 | 275 | 70 | ||
| GenBank mRNA | AF068058.1 | - | 1 - 2 | 211 | 70 | |
| AF088887.1 | - | 1 - 2 | 165 | 70 | ||
| DQ304490.1 | - | 1 - 2 | 165 | 70 | ||
| KT216058.1 | - | 1 - 2 | 165 | 70 | ||
More Information
Set Membership: |
|

IL10
Genomic Map