Assay Details
| Gene Symbol: BSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082047.1 | NP_001075516.1 | 1785 | 80 | ||
| GenBank mRNA | AF109472.1 | - | 1785 | 80 | ||
| Gene Symbol: LOC103344941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008253072.1 | XP_008251294.1 | 631 | 80 | ||
More Information
Set Membership: |
|

BSP1
Genomic Map