Assay Details
| Gene Symbol: KLF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082053.1 | NP_001075522.1 | 1 - 2 | 428 | 88 | |
| XM_008259978.2 | XP_008258200.1 | 2 - 3 | 1456 | 88 | ||
| GenBank mRNA | AF131882.1 | - | 1 - 2 | 428 | 88 | |
More Information
Set Membership: |
|

KLF5
Genomic Map