Assay Details
| Gene Symbol: PPIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082057.1 | NP_001075526.1 | 1 | 422 | 61 | |
| GenBank mRNA | AF139893.1 | - | 1 | 422 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPIA| Gene Symbol: PPIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082057.1 | NP_001075526.1 | 1 | 422 | 61 | |
| GenBank mRNA | AF139893.1 | - | 1 | 422 | 61 | |
Set Membership: |
|