Assay Details
| Gene Symbol: CA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082086.1 | NP_001075555.1 | 4 - 5 | 431 | 75 | |
| XM_017347899.1 | XP_017203388.1 | 4 - 5 | 567 | 75 | ||
| GenBank mRNA | AF263367.2 | - | 4 - 5 | 431 | 75 | |
More Information
Set Membership: |
|

CA12
Genomic Map