Assay Details
| Gene Symbol: EPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082090.1 | NP_001075559.1 | 1 - 2 | 162 | 86 | |
| GenBank mRNA | AF290943.1 | - | 1 - 2 | 162 | 86 | |
More Information
Set Membership: |
|

EPO
Genomic Map