Assay Details
| Gene Symbol: LIPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082106.1 | NP_001075575.1 | 7 - 8 | 1079 | 111 | |
| XM_008266161.2 | XP_008264383.1 | 7 - 8 | 1091 | 111 | ||
| GenBank mRNA | AF351188.1 | - | 7 - 8 | 1079 | 111 | |
More Information
Set Membership: |
|

LIPH
Genomic Map