Assay Details
| Gene Symbol: DBI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082113.1 | NP_001075582.1 | 2 - 3 | 177 | 55 | |
| GenBank mRNA | AF407578.2 | - | 2 - 3 | 177 | 55 | |
More Information
Set Membership: |
|

DBI
Genomic Map