Assay Details
| Gene Symbol: AK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082114.1 | NP_001075583.1 | 4 - 5 | 567 | 121 | |
| GenBank mRNA | AF417508.1 | - | 4 - 5 | 567 | 121 | |
More Information
Set Membership: |
|

AK3
Genomic Map