Assay Details
| Gene Symbol: SLC22A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082115.1 | NP_001075584.1 | 3 - 4 | 711 | 68 | |
| GenBank mRNA | AF458095.1 | - | 3 - 4 | 711 | 68 | |
More Information
Set Membership: |
|

SLC22A2
Genomic Map