Assay Details
| Gene Symbol: ANXA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082119.1 | NP_001075588.1 | 9 - 10 | 782 | 62 | |
| XM_008255642.2 | XP_008253864.1 | 8 - 9 | 735 | 62 | ||
| GenBank mRNA | AF510726.1 | - | 9 - 10 | 782 | 62 | |
More Information
Set Membership: |
|

ANXA13
Genomic Map