Assay Details
| Gene Symbol: PCBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082124.1 | NP_001075593.1 | 1 | 555 | 72 | |
| GenBank mRNA | AJ003023.1 | - | 1 | 555 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCBP1| Gene Symbol: PCBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082124.1 | NP_001075593.1 | 1 | 555 | 72 | |
| GenBank mRNA | AJ003023.1 | - | 1 | 555 | 72 | |
Set Membership: |
|