Assay Details
| Gene Symbol: GABARAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082142.1 | NP_001075611.1 | 3 - 4 | 318 | 129 | |
| GenBank mRNA | AY094174.1 | - | 3 - 4 | 318 | 129 | |
More Information
Set Membership: |
|

GABARAP
Genomic Map