Assay Details
| Gene Symbol: FKBP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082145.1 | NP_001075614.1 | 2 - 3 | 199 | 101 | |
| GenBank mRNA | AY159324.1 | - | 2 - 3 | 199 | 101 | |
More Information
Set Membership: |
|

FKBP1B
Genomic Map