Assay Details
| Gene Symbol: PDZK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082151.2 | NP_001075620.1 | 3 | 348 | 71 | |
| XM_008263954.2 | XP_008262176.1 | 4 | 478 | 71 | ||
| XM_008263955.2 | XP_008262177.1 | 3 | 527 | 71 | ||
| XM_008263956.2 | XP_008262178.1 | 3 | 328 | 71 | ||
| GenBank mRNA | AY204473.1 | - | 3 | 351 | 71 | |
More Information
Set Membership: |
|

PDZK1
Genomic Map