Assay Details
| Gene Symbol: PAPSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082173.1 | NP_001075642.1 | 10 - 11 | 1473 | 72 | |
| GenBank mRNA | AY618932.1 | - | 10 - 11 | 1473 | 72 | |
More Information
Set Membership: |
|

PAPSS2
Genomic Map