Assay Details
| Gene Symbol: LCAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082190.1 | NP_001075659.1 | 1 - 2 | 161 | 70 | |
| GenBank mRNA | D13668.1 | - | 1 - 2 | 161 | 70 | |
More Information
Set Membership: |
|

LCAT
Genomic Map