Assay Details
| Gene Symbol: GC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082204.1 | NP_001075673.1 | 6 - 7 | 715 | 67 | |
| GenBank mRNA | D29666.1 | - | 6 - 7 | 715 | 67 | |
More Information
Set Membership: |
|

GC
Genomic Map