Assay Details
| Gene Symbol: PRLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082231.1 | NP_001075700.1 | 7 - 8 | 1161 | 70 | |
| XM_017344477.1 | XP_017199966.1 | 5 - 6 | 775 | 70 | ||
| XM_017344478.1 | XP_017199967.1 | 8 - 9 | 1414 | 70 | ||
| XM_017344480.1 | XP_017199969.1 | 9 - 10 | 1464 | 70 | ||
| XM_017344481.1 | XP_017199970.1 | 6 - 7 | 807 | 70 | ||
| XM_017344482.1 | XP_017199971.1 | 7 - 8 | 812 | 70 | ||
| XM_017344483.1 | XP_017199972.1 | 7 - 8 | 1349 | 70 | ||
| XM_017344484.1 | XP_017199973.1 | 6 - 7 | 1315 | 70 | ||
| GenBank mRNA | J04510.1 | - | 7 - 8 | 1161 | 70 | |
More Information
Set Membership: |
|

PRLR
Genomic Map