Assay Details
| Gene Symbol: TIMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082232.2 | NP_001075701.2 | 2 - 3 | 170 | 85 | |
| XM_008272340.2 | XP_008270562.1 | 2 - 3 | 281 | 85 | ||
| GenBank mRNA | AY829730.1 | - | 1 - 2 | 114 | 85 | |
| J04712.1 | - | 1 - 2 | 141 | 85 | ||
More Information
Set Membership: |
|

TIMP1
Genomic Map