Assay Details
| Gene Symbol: CALR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082235.1 | NP_001075704.1 | 1024 | 62 | ||
| GenBank mRNA | J05138.1 | - | 1024 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CALR| Gene Symbol: CALR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082235.1 | NP_001075704.1 | 1024 | 62 | ||
| GenBank mRNA | J05138.1 | - | 1024 | 62 | ||
Set Membership: |
|