Assay Details
| Gene Symbol: SCGB1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082237.1 | NP_001075706.1 | 288 | 68 | ||
| GenBank mRNA | K01657.1 | - | 288 | 68 | ||
| M27564.1 | - | 178 | 68 | |||
More Information
Set Membership: |
|

SCGB1A1
Genomic Map