Assay Details
| Gene Symbol: MSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082248.1 | NP_001075717.1 | 2 - 3 | 237 | 89 | |
| GenBank mRNA | D13381.1 | - | 2 - 3 | 221 | 89 | |
| L11692.1 | - | 2 - 3 | 237 | 89 | ||
| L11693.1 | - | 2 - 3 | 237 | 89 | ||
More Information
Set Membership: |
|

MSR1
Genomic Map