Assay Details
| Gene Symbol: GSTM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082252.1 | NP_001075721.1 | 4 - 5 | 297 | 69 | |
| GenBank mRNA | L23766.1 | - | 4 - 5 | 297 | 69 | |
| Gene Symbol: LOC100356307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715794.3 | XP_002715840.3 | 4 - 5 | 339 | 69 | |
| Gene Symbol: LOC100357148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715796.3 | XP_002715842.1 | 4 - 5 | 335 | 69 | |
More Information
Set Membership: |
|

GSTM2
Genomic Map